Ethyl-3-piperidinecarboxylate

Supplier: Molekula

Synonyms: Ethyl piperidine-3-carboxylateEthyl nipecotate

MOLE46246962-100GEA 66.7 EUR
MOLE46246962-100G
Ethyl-3-piperidinecarboxylate
Ethyl 3-piperidinecarboxylate
Formula: C₈H₁₅NO₂
MW: 157,21 g/mol
MDL Number: MFCD00005991
CAS Number: 5006-62-2
EINECS: 225-681-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR