Galvanised iron zinc coated, wire gauzes with ceramic center for gas burners
Supplier: MARIENFELD
MARI6613101EA 45.1 EUR
MARI6613101 MARI6613105 MARI6613103
Galvanised iron zinc coated, wire gauzes with ceramic center for gas burners
Galvanised iron zinc coated
These wire gauzes with ceramic centre are corrosion resistant thanks to their zinc coating.
  • Asbestos-free
  • No hazardous emission when being used
  • Considered harmless for environment