pH meter, handheld, ad 110 pH
Supplier: Amarell
AMARE918000EA 75.3 EUR
AMARE918000
pH meter, handheld, ad 110 pH
Electrochemical Meters pH and Multi-Parameter Meters
Easy-to-use pH meter for universal use.
  • With screwed-on electrode (interchangeable)
  • Can be kept safe in the practical storage case
  • Battery life of approx. 1000 hours
Delivery information: Supplied with interchangeable pH electrode and calibration set.