Syringe filters, Acrodisc® with Supor® membrane
Supplier: Pall Laboratory
514-0861EA 237 EUR
514-0861 514-0862 514-1105
Syringe filters, Acrodisc® with Supor® membrane
Filters Syringe Filters
These filters have a Supor® hydrophilic polyethersulfone (PES) membrane, PP housing and borosilicate glass GxF prefilter. Universal life science membrane for general aqueous samples.
  • Low protein binding to minimise sample loss
  • Built-in prefilter for increased throughput of difficult to filter liquids
  • Used in dissolution testing