Water purification system, Select Purewater 300
Supplier: Purite
PURIL300475EA 9010 EUR
PURIL300475 171-0882
Water purification system, Select Purewater 300
Water Purification Systems Pure and Ultra Pure Water Systems
The Select Purewater 300 is designed specifically for use with laboratory glassware washing machines, providing rinse water to a purity of over 1 MΩhm.cm at a flow rate of up to 48 litres per hour. The system is compact, robust, simple to use and easy to maintain, using proven pre-treatment, reverse osmosis and ion-exchange deionisation technology to ensure a reliable and consistent supply of purified water.
  • High quality reverse osmosis and pre-treatment modules ensure a reliable and consistent quality of water
  • Water quality is constantly monitored and displayed
  • Compact footprint, saving on valuable bench space
  • Minimal external connections for easy installation
  • System information is easily accessible
  • Alarms notify the user of changes
The unit has standard connections for quick installation, with a high flow outlet that fits easily to most glassware washing machines using a conventional gravity feed. It incorporates a standard 20 litre integral tank to connect to a lab washing machine (optional external tanks available for extra capacity).
The Purewater 300 can be fitted with an on-demand pure water boost pump to provide up to 1,5 bar pressurised feed with a flow rate of up to 200 litres per hour.