Magnetic hotplate stirrer, analogue, HSC
Supplier: VELP Scientifica
VELPF20500101EA 339 EUR
VELPF20500101
Magnetic hotplate stirrer, analogue, HSC
Hot plate stirrer with a white ceramic heating plate which is easy to clean. The unit has a rugged low profile design, a die cast body with an inclined control panel giving easy access and visibility. The HSC features a run off groove which provides protection from liquid spills, the square shaped ceramic heating plate resists acids, bases and solvents.
  • Technopolymer casing resists chemicals, scratches and surface abrasions
  • White ceramic plate is ideal for observing colour change titrations
  • Metal vessels can be used on the ceramic plate
  • Control panel is separated from the heating plate providing enhanced safety
  • High-power PCM type magnet with mono-phase motor for continuous operation
IP protection class according to DIN EN 60529: IP 42