You Searched For: Electric Burners
The electric burners combine the advantages of a regular gas burner with the simple operation of a heating mantle. While the heating element guides the radiant heat to the top cavity, the top cowl keeps the extreme temperatures safely away from users. Electric burners can be used for heating test tubes, crucibles, small flasks, and beakers. Air circulation holes allow the unit to be held by the base or placed on a bench top while operating.
The electric burners combine the advantages of a regular gas burner with the simple operation of a heating mantle. While the heating element guides the radiant heat to the top cavity, the top cowl keeps the extreme temperatures safely away from users. Electric burners can be used for heating test tubes, crucibles, small flasks, and beakers. Air circulation holes allow the unit to be held by the base or placed on a bench top while operating.
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (HORO7001-7002-7003)
Supplier: HORO DR ING HOFMAN GMBH
Description: Electric laboratory burner which can be operated as a 'stand alone' burner or with a glass support and/or power regulator.
UOM: 1 * 1 SET
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.