You Searched For: Preserved Specimens

Preserved specimens are delivered in sealed containers for long-lasting viability in transit, and arrive ready to be used for dissection purposes. Whole organisms or discrete organs are available, with skinned specimens all maintaining intact and undamaged internal organs. High-quality preserved specimens are packed in a variety of containers depending on size and intended use, and a vast selection of species sourced from the land, air, and sea are available for clinical or classroom dissection.

Preserved specimens are delivered in sealed containers for long-lasting viability in transit, and arrive ready to be used for dissection purposes. Whole organisms or discrete organs are available, with skinned specimens all maintaining intact and undamaged internal organs. High-quality preserved specimens are packed in a variety of containers depending on size and intended use, and a vast selection of species sourced from the land, air, and sea are available for clinical or classroom dissection.


6  results were found

SearchResultCount:"6"

Sort Results

List View Easy View

Rate These Search Results

Catalog Number: (ALEXADILYTRB100F)
Supplier: Enzo Life Sciences
Description: Rat brain tissue extract
UOM: 1 * 200 µG


Catalog Number: (ALEXADILYTHM100F)
Supplier: Enzo Life Sciences
Description: Microsomes are a rich source of heme-containing metabolic enzymes, including heme-oxygenases and cytochrome P450 (CYP) enzymes, and are commonly used ...
UOM: 1 * 200 µG


Catalog Number: (ALEXBML-SW106-0200)
Supplier: Enzo Life Sciences
Description: Whole liver extract from mouse useful as a Western blotting control for Aryl-hydrocarbon receptor antibodies or for use with other antibodies/proteins...
UOM: 1 * 200 µl


Catalog Number: (ALEXBML-SW115-0100)
Supplier: Enzo Life Sciences
Description: Whole cell extract of human Jurkat macrophage cells.
UOM: 1 * 100 µl


Catalog Number: (ALEXBML-SW110-0200)
Supplier: Enzo Life Sciences
Description: Whole thymus extract from mouse designed for Western blotting control of anti-PARP antibodies or other antibodies/proteins.
UOM: 1 * 200 µl


Catalog Number: (ICNA092850145)
Supplier: MP Biomedicals
Description: Chicken embryo extract prepared from nine to 12 day old eggs.
UOM: 1 * 20 mL

MSDS


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 6 of 6
no targeter for Bottom