Oxygen meter, handheld, GOX 100T
Supplier: Greisinger
GRSG600157EA 276 EUR
GRSG600157
Oxygen meter, handheld, GOX 100T
This portable device precisely measures the oxygen concentration in gases. Ideal for diving applications, e.g. for measuring the oxygen concentration in oxygen bottles.
  • One-button calibration
  • MOD-Display (Maximum Operating Depth)
  • HOLD function
  • Automatic power-off (selectable: 1 to 120 min)
  • Durable ABS housing, front side IP 65
Please note: The meter can not be used under water!
Delivery information: Meter is supplied with sensor GOEL 370, T-piece, flow diverter, battery and manual.