Chemistry vacuum systems, model 3012 NT, VARIO®
Supplier: Vacuubrand
VACU20743800EA 14530 EUR
VACU20743800 VACU20743700 VACU20743801
Chemistry vacuum systems, model 3012 NT, VARIO®
These VARIO® pumping units feature a very high pumping speed even close to the outstanding ultimate vacuum and are ideal for high vacuum requirements with most high boiling solvents. They provide precise vacuum control by continuously adjusting the pump´s motor speed. The controller provides fully automatic evaporations without any need for parameter input. The pump design offers exceptionally high chemical resistance supporting almost universal usage in chemistry and pharmaceutics.
  • Automatic adjustment of the vacuum level throughout the process for high process reproducibility and unattended operation
  • Short process times due to zero-fluctuation (hysteresis-free) vacuum control, even for large amounts of vapor
  • Removable CVC 3000 vacuum controller, can be arranged flexibly
  • Extraordinary diaphragm life for minimum operational and servicing costs
The PC 3012 NT VARIO pumping unit relies on a well-proven operating concept for evaporations with large amounts of solvents. The inlet separator (AK) retains particles and liquid droplets, the waste vapor condenser at the outlet (EK) is highly efficient and compact. The condenser enables near-100% solvent recovery, efficient recycling and active protection of the environment.
The high pumping speed reduces the process time and meets the high vacuum requirements, for example, of parallel operations in vacuum networks. Evaporation processes can be run fully automatically and with short process time without sacrificing sensitivity. The VARIO® control minimizes the danger of superheating and foaming and therefore ensures consistent process safety and protection of samples. With its inlet separator and efficient solvent recovery system, the PC 3012 NT VARIO is ideal for rough operating conditions.