Ammonia-D3 solution 25% in D₂O (99.5% D), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

8.15008.0010 8.15008.0025
8.15008.0010EA 144 EUR
8.15008.0010 8.15008.0025
Ammonia-D3 solution 25% in D₂O (99.5% D), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Ammonia-D3
Formula: H₅NO
MW: 40,077 g/mol
MDL Number: MFCD00064511
CAS Number: 12168-30-8
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Solubility (20 °C) soluble
Boiling Point 36 °C
Vapor Pressure 535 hPa (20 °C)
Density 1.056 g/cm3 (20 °C)
pH (H2O, 20 °C) strongly alkaline
Hazard Class 8
Hazard Class Description Corrosive Material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR