You Searched For: Contact Plates for Microbiology

Contact plates efficiently monitor environmental surfaces for microbial contamination. Used to determine the sanitation techniques and schedules required, the plates enumerate the colonies collected before and after cleaning is completed. With a marked grid on the flat bottom, optical counting can be easily done. Already containing the media and supplements, plates will deactivate any disinfectant residue. The contact plates allow critical cleanroom laboratories to meet and maintain sterile conditions for susceptible tests.

Contact plates efficiently monitor environmental surfaces for microbial contamination. Used to determine the sanitation techniques and schedules required, the plates enumerate the colonies collected before and after cleaning is completed. With a marked grid on the flat bottom, optical counting can be easily done. Already containing the media and supplements, plates will deactivate any disinfectant residue. The contact plates allow critical cleanroom laboratories to meet and maintain sterile conditions for susceptible tests.


20  results were found

SearchResultCount:"20"

Sort Results

List View Easy View

Rate These Search Results

Supplier: VWR Chemicals
Description: Available in two pack sizes - triple-wrapped (24 plate pack) and double-wrapped (30 plate pack). Plates are packed in a pack with six blisters. Each b...
Supplier: VWR Chemicals
Description: The range is manufactured from high quality raw materials. All culture media are manufactured using selected dehydrated bases, and comply with the req...
Supplier: VWR Chemicals
Description: Contact plates are ideal for monitoring microbiological contamination of surfaces and air (using an air sampler) inside cleanrooms, isolators, RABS (R...
Supplier: VWR Chemicals
Description: These contact plates, with triple-bagged sterile packaging and lockable lid, are ideal for environmental control of air, surfaces, gloves and overalls...

MSDS Certificates New Product Sale

Catalog Number: (710-0961)
Supplier: VWR Collection
Description: Used in combination with contact plates, the contact weight enables the standardisation of microbiological surface control between operators. It appli...
UOM: 1 * 1 items


Supplier: LIOFILCHEM S.R.L.
Description: The peelable blister pack fulfills the highest quality and protection standards for the plate. The blister pack, produced fully automatically and in c...

Supplier: LIOFILCHEM S.R.L.
Description: The new peelable blister pack fulfills the highest quality and protection standards for the plate. The blister pack, produced fully automatically and ...

Supplier: 3M Food Safety
Description: 3Mâ„¢ Petrifilmâ„¢ Rapid E. coli/coliform count plates provide a cost-effective, convenient and reliable method for testing equipment, raw material...

Supplier: THERMO FISHER DIAGNOSTICS GMBH
Description: Contact plate, Sabouraud dextrose, triple wrapped, Application: Yeast and moulds

Supplier: 3M Food Safety
Description: The 3Mâ„¢ Petrifilmâ„¢ High-sensitivity coliform count plate accommodates 5 ml samples and detection of low levels of confirmed coliforms in 24 hours.

Supplier: 3M Food Safety
Description: 3Mâ„¢ Petrifilmâ„¢ Rapid coliform count plates are designed to provide fast, accurate results in detecting coliform contamination.

Supplier: 3M Food Safety
Description: 3Mâ„¢ Petrifilmâ„¢ Rapid Yeast and Mold Count Plates simplify and accelerate detection. Featuring a new indicator technology that makes colonies easier to...
Supplier: 3M Food Safety
Description: The 3Mâ„¢ Petrifilmâ„¢ Lactic acid bacteria count plate is a time-saving, sample-ready plate designed to determine total lactic acid bacteria populations ...
Supplier: NISSUI PHARMA SOLUTIONS
Description: Ready to use dehydrated film media in rigid plastic with lid for cultivating and counting microorganisms in food, for monitoring product quality and p...

Catalog Number: (APPC433745.0922)
Supplier: PanReac AppliChem
Description: Violet red bile glucose agar (VRBG) is used as a culture media for microbiology.
UOM: 1 * 30 items


Catalog Number: (SCIL1006731)
Supplier: HYSERVE GMBH & CO. KG
Description: Ready to use dehydrated film media in rigid plastic with lid for cultivating and counting microorganisms in food, for monitoring product quality and p...
UOM: 1 * 240 items


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 16 of 20
no targeter for Bottom