Anti-BMPER Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-6910REA 691 EUR
BOSSBS-6910R
Anti-BMPER Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BMPER
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR