Anti-FBXO2 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP10212BEA 348 EUR
ABGEAP10212B
Anti-FBXO2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FBXO2 (F-box protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR