Anti-SMAD2 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP3654AEA 348 EUR
ABGEAP3654A
Anti-SMAD2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SMAD2 (SMAD family member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR