Anti-ULK2 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT4464AEA 350 EUR
ABGEAT4464A
Anti-ULK2 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: ULK2 (unc-51 like autophagy activating kinase 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR