Anti-UNC13D Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT4470AEA 350 EUR
ABGEAT4470A
Anti-UNC13D Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: UNC13D (unc-13 homolog D (C. elegans))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR