Anti-USP4 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT4490AEA 350 EUR
ABGEAT4490A
Anti-USP4 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: USP4 (ubiquitin specific peptidase 4 (proto-oncogene))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR