Anti-VPREB3 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT4517AEA 350 EUR
ABGEAT4517A
Anti-VPREB3 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: VPREB3 (pre-B lymphocyte 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR