Anti-VPS11 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT4518AEA 350 EUR
ABGEAT4518A
Anti-VPS11 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: VPS11 (vacuolar protein sorting 11 homolog (S. cerevisiae))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR