Anti-ZNF37A Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT4630AEA 350 EUR
ABGEAT4630A
Anti-ZNF37A Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: ZNF37A (zinc finger protein 37A)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR