Anti-Ly-6G Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 1A8]

Supplier: Tonbo Biosciences
20-1276-U025EA 43.6 EUR
20-1276-U025 20-1276-U100
Anti-Ly-6G Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 1A8]
Antibodies
The 1A8 antibody binds to mouse Ly-6G, commonly known as Gr-1, a member of the Ly-6 superfamily of GPI-anchored cell surface proteins with roles in cell signaling and cell adhesion. Gr-1 is differentially expressed during development and maturation of cells in the myeloid lineage and is expression at varying stages and levels on monocytes, macrophages, granulocytes, and peripheral neutrophils.

Type: Primary
Antigen: Ly-6G (Ly-6G)
Clonality: monoclonal
Clone: 1A8
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR