Anti-FCER1 Armenian Hamster Monoclonal Antibody (FITC (Fluorescein)) [clone: MAR-1]

Supplier: Tonbo Biosciences
35-5898-U025EA 31.6 EUR
35-5898-U025 35-5898-U100
Anti-FCER1 Armenian Hamster Monoclonal Antibody (FITC (Fluorescein)) [clone: MAR-1]
Antibodies
The MAR-1 antibody reacts with the Fc epsilon Receptor I alpha chain (FceRIa), a transmembrane protein member of the Ig superfamily. This chain, together with a beta chain and two gamma chains form a tetrameric complex that supports IgE-mediated signaling and subsequent release of chemical mediators of allergy and immediate hypersensitivity. FceR1a is upregulated in the presence of IgE on those cell types which express it, such as Mast cells and Basophils.

Type: Primary
Antigen: FCER1 (Fc fragment of IgE, high affinity I, receptor for)
Clonality: monoclonal
Clone: 42064
Conjugation: FITC (Fluorescein)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR