Anti-CD45RO Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: UCHL1]

Supplier: Tonbo Biosciences
50-0457-T025EA 41.8 EUR
50-0457-T025 50-0457-T100
Anti-CD45RO Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: UCHL1]
Antibodies
The UCHL1 antibody reacts with the human CD45 isoform known as CD45RO, a protein tyrosine phosphatase of 220 kDa. CD45 is one of the most abundant hematopoietic markers, and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). Various isoforms are generated and expressed in cell-specific patterns. With their broad cell distribution, CD45 isoforms are critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RA and CD45RB.

Type: Primary
Antigen: CD45RO (CD45RO)
Clonality: monoclonal
Clone: UCHL1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR