Anti-CD45.1 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: A20]

Supplier: Tonbo Biosciences
60-0453-U025EA 35.3 EUR
60-0453-U025 60-0453-U100
Anti-CD45.1 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: A20]
Antibodies
The A20 antibody reacts with mouse CD45.1, also known as Ly5.1, which is a strain-specific allelic form of the CD45 Leukocyte Common Antigen (LCA). Functionally, CD45 is a protein tyrosine phosphatase whose broad cell distribution supports a critical role in many leukocyte functions, including regulation of signal transduction and cell activation associated with the T cell and B cell receptors.

Type: Primary
Antigen: CD45.1 (CD45.1)
Clonality: monoclonal
Clone: A20
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR