Anti-CD44 Rat Monoclonal Antibody (violetFluor® 450) [clone: IM7]

Supplier: Tonbo Biosciences
75-0441-U100EA 147 EUR
75-0441-U100 75-0441-U025
Anti-CD44 Rat Monoclonal Antibody (violetFluor® 450) [clone: IM7]
Antibodies
The IM7 antibody recognizes CD44, a ubiquitously expressed cell surface receptor which is important for extracellular matrix organization, cell-cell and cell-matrix adhesion and migration. CD44 may be expressed in a number of different isoforms (splice variants) from the most typical or “standard” form, known as CD44s, to variants designated CD44v, e.g. CD44v1 or CD44v6. These receptors interact with several ligands, but most often associate with an extracellular matrix component hyaluronate, through which it mediates adhesion.

Type: Primary
Antigen: CD44 (CD44 molecule (Indian blood group))
Clonality: monoclonal
Clone: IM7
Conjugation: violetFluor® 450
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR