Anti-DDK Tag Antibody Agarose beads

Supplier: OriGene
ORIGTA150038EA 1490 EUR
ORIGTA150038
Anti-DDK Tag Antibody Agarose beads
Type: Tag
Antigen: DDK Tag
Clonality:
Clone:
Conjugation: Agarose beads
Epitope:
Host: None
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR