Anti-tGFP Mouse Antibody (Magnetic beads)

Supplier: OriGene
ORIGTA150039EA 1050 EUR
ORIGTA150039
Anti-tGFP Mouse Antibody (Magnetic beads)
Antibodies
Type: Tag
Antigen: tGFP
Clonality:
Clone:
Conjugation: Magnetic beads
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR