Anti-tGFP Mouse Monoclonal Antibody

Supplier: OriGene
ORIGTA150041EA 450 EUR
ORIGTA150041
Anti-tGFP Mouse Monoclonal Antibody
Antibodies
Type: Tag
Antigen: tGFP
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR