Anti-MED20 Mouse Monoclonal Antibody

Supplier: OriGene
ORIGTA150061EA 497 EUR
ORIGTA150061
Anti-MED20 Mouse Monoclonal Antibody
Antibodies
Type: Tag
Antigen: MED20 (mediator complex subunit 20)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR