Anti-mRFP Mouse Monoclonal Antibody

Supplier: OriGene
ORIGTA180093EA 497 EUR
ORIGTA180093
Anti-mRFP Mouse Monoclonal Antibody
Antibodies
Type: Tag
Antigen: mRFP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR