Anti-PPP2CA Rabbit monoclonal antibody

ORIGTA300307EA 395 EUR
ORIGTA300307
Anti-PPP2CA Rabbit monoclonal antibody
Type: Primary
Antigen: PPP2CA (protein phosphatase 2, catalytic subunit, alpha isozyme)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR