Anti-EIF2AK2 Rabbit monoclonal antibody [clone: YE350]

Supplier: OriGene
ORIGTA300449EA 685 EUR
ORIGTA300449
Anti-EIF2AK2 Rabbit monoclonal antibody [clone: YE350]
Recommended Dilutions: Western Blot: 1: 1000-5000; Immunochemistry: 1:250-500; Immunoprecipitation: 1: 100

Type: Primary
Antigen: EIF2AK2 (eukaryotic translation initiation factor 2-alpha kinase 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR