Anti-CALM2 Rabbit Monoclonal Antibody [clone: EP799Y]

Supplier: OriGene
ORIGTA300636EA 792 EUR
ORIGTA300636
Anti-CALM2 Rabbit Monoclonal Antibody [clone: EP799Y]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-1:5000; Immunochemistry/Immunofluorescence: 1:100-1:250; Flow Cytometry: 1:50; Immunoprecipitation: 1:50

Type: Primary
Antigen: CALM2 (calmodulin 2 (phosphorylase kinase, delta))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR