Anti-MYB Rabbit Monoclonal Antibody [clone: EP769Y]

Supplier: OriGene
ORIGTA300706EA 679 EUR
ORIGTA300706
Anti-MYB Rabbit Monoclonal Antibody [clone: EP769Y]
Recommended Dilutions: Western Blot: 1:1000-1:5000; Immunochemistry/Immunofluorescence: 1:100-1:250; Immunoprecipitation: 1:50

Type: Primary
Antigen: MYB (v-myb avian myeloblastosis viral oncogene homolog)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR