Anti-PRNP Rabbit Monoclonal Antibody [clone: EP1802Y]

Supplier: OriGene
ORIGTA300958EA 699 EUR
ORIGTA300958
Anti-PRNP Rabbit Monoclonal Antibody [clone: EP1802Y]
Antibodies
Recommended Dilutions: Western Blot: 1:5000-1:10000; Flow Cytometry: 1:200; Immunochemistry/Immunofluorescence: 1:100-1:250

Type: Primary
Antigen: PRNP (prion protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR