Anti-RARA Rabbit monoclonal antibody

ORIGTA300976EA 443 EUR
ORIGTA300976
Anti-RARA Rabbit monoclonal antibody
Recommended Dilutions: Western Blot: 1:500-1:1000; Flow Cytometry: 1:50

Type: Primary
Antigen: RARA (retinoic acid receptor, alpha)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR