Anti-CASR Mouse Monoclonal Antibody [clone: HL 1499]

Supplier: OriGene
ORIGTA301411EA 658 EUR
ORIGTA301411
Anti-CASR Mouse Monoclonal Antibody [clone: HL 1499]
Antibodies
Recommended Dilutions: Western Blot: 1:50-1:200

Type: Primary
Antigen: CASR (calcium-sensing receptor)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR