Anti-NQO1 Mouse Monoclonal Antibody [clone: A180]

Supplier: OriGene
ORIGTA301418EA 590 EUR
ORIGTA301418
Anti-NQO1 Mouse Monoclonal Antibody [clone: A180]
Antibodies
Recommended Dilutions: Western Blot: 1: 1000-1: 10000

Type: Primary
Antigen: NQO1 (NAD(P)H dehydrogenase, quinone 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR