Anti-HSPB1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA301440EA 516 EUR
ORIGTA301440
Anti-HSPB1 Rabbit Polyclonal Antibody
Recommended Dilutions: Western Blot: 0.5-1.0 ?g/ml

Type: Primary
Antigen: HSPB1 (heat shock 27kDa protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR