Anti-CAV1 Mouse Monoclonal Antibody [clone: 7C8]

Supplier: OriGene
ORIGTA301445EA 650 EUR
ORIGTA301445
Anti-CAV1 Mouse Monoclonal Antibody [clone: 7C8]
Antibodies
Recommended Dilutions: Western Blot: 1: 1000-1:4000

Type: Primary
Antigen: CAV1 (caveolin 1, caveolae protein, 22kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR