Anti-PDK1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA301454EA 624 EUR
ORIGTA301454
Anti-PDK1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 2?g/ml

Type: Primary
Antigen: PDK1 (pyruvate dehydrogenase kinase, isozyme 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR