Anti-SAT1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA301460EA 711 EUR
ORIGTA301460
Anti-SAT1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: SAT1 (spermidine/spermine N1-acetyltransferase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR