Anti-FANCG Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA301479EA 516 EUR
ORIGTA301479
Anti-FANCG Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: FANCG (Fanconi anemia, complementation group G)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR