Anti-BAP1 Mouse Monoclonal Antibody [clone: 1G8]

Supplier: OriGene
ORIGTA301480EA 545 EUR
ORIGTA301480
Anti-BAP1 Mouse Monoclonal Antibody [clone: 1G8]
Antibodies
Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: BAP1 (BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR