Anti-ATG5 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA301481EA 747 EUR
ORIGTA301481
Anti-ATG5 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: ATG5 (autophagy related 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR