Anti-XRCC3 Mouse Monoclonal Antibody [clone: 10F1/6]

Supplier: OriGene
ORIGTA301487EA 509 EUR
ORIGTA301487
Anti-XRCC3 Mouse Monoclonal Antibody [clone: 10F1/6]
Antibodies
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: XRCC3 (X-ray repair complementing defective repair in Chinese hamster cells 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR