Anti-NKX3-1 Mouse Monoclonal Antibody

Supplier: OriGene
ORIGTA301499EA 620 EUR
ORIGTA301499
Anti-NKX3-1 Mouse Monoclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: NKX3-1 (NK3 homeobox 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR