Anti-PARK7 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA301508EA 658 EUR
ORIGTA301508
Anti-PARK7 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PARK7 (parkinson protein 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR