Anti-DNM1L Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA301510EA 590 EUR
ORIGTA301510
Anti-DNM1L Rabbit Polyclonal Antibody
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: DNM1L (dynamin 1-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR